Anhydrid kyseliny trifluoroctové 99%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
Trifluoracetic anhydride, TFAA
PC7170-500G
PC7170-1KG
PC7170-5KG
APOSPC7170-500GEA
1660
CZK
APOSPC7170-500G
APOSPC7170-1KG
APOSPC7170-5KG
Anhydrid kyseliny trifluoroctové 99%
Anhydrid kyseliny trifluoroctové
Useful trifluoroacetylating agent for the protectionof alcohols & amines.
Vzorec:
C₄F₆O₃ MW: 210,03 g/mol Bod varu: 39…40 °C Bod tání: –65 °C Hustota: 1,511 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000416 CAS číslo: 407-25-0 EINECS: 206-982-9 UN: 3265 ADR: 8,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...