Tisk…

3-Fluoro-2,4-dimethoxyaniline 98%

Dodavatel: Apollo Scientific

Synonyms: 2,4-Dimethoxy-3-fluoroaniline

PC7135-1G PC7135-5G
APOSPC7135-1GEA 10820 CZK
APOSPC7135-1G APOSPC7135-5G
3-Fluoro-2,4-dimethoxyaniline 98%
3-Fluoro-2,4-dimethoxyaniline
Vzorec: C₈H₁₀FNO₂
Storage Temperature: Ambient
MDL Number: MFCD09258660
CAS číslo: 195136-66-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR