3-Fluoro-4-(2,2,2-trifluoroethoxy)phenylboronic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(3-Fluoro-4-(2,2,2-trifluoroethoxy)phenyl)boronic acid
PC7134-1G
PC7134-250MG
APOSPC7134-1GEA
12440
CZK
APOSPC7134-1G
APOSPC7134-250MG
3-Fluoro-4-(2,2,2-trifluoroethoxy)phenylboronic acid 98%
3-Fluoro-4-(2,2,2-trifluoroethoxy)phenylboronic acid
Vzorec:
C₈H₇BF₄O₃ MW: 237,9400 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD09258747 CAS číslo: 947533-09-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...