Tisk…

2-Adamantyl-4-bromo-1-(2,2,2-trifluoroethoxy)benzene 98%

Dodavatel: Apollo Scientific

Synonyms: 1-[5-Bromo-2-(2,2,2-trifluoroethoxy)phenyl]adamantane

PC7133-1G PC7133-250MG
APOSPC7133-1GEA 11210 CZK
APOSPC7133-1G APOSPC7133-250MG
2-Adamantyl-4-bromo-1-(2,2,2-trifluoroethoxy)benzene 98%
2-Adamantyl-4-bromo-1-(2,2,2-trifluoroethoxy)benzene
Vzorec: C₁₈H₂₀BrF₃O
Storage Temperature: Ambient
MDL Number: MFCD09258647
CAS číslo: 929000-50-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR