N-Boc-3,5-difluoro-L-phenylalanine 99%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Boc-Phe(3,5-DiF)-OH
PC7119-1G
PC7119-250MG
PC7119-5G
APOSPC7119-1GEA
1080
CZK
APOSPC7119-1G
APOSPC7119-250MG
APOSPC7119-5G
N-Boc-3,5-difluoro-L-phenylalanine 99%
N-Boc-3,5-difluoro-L-phenylalanine
Vzorec:
C₁₄H₁₇F₂NO₄ MW: 301,29 g/mol Bod tání: 115 °C Storage Temperature: Ambient |
MDL Number:
MFCD00797555 CAS číslo: 205445-52-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...