Triethyloxonium tetrafluoroborate 95%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC7110-100G
PC7110-25G
APOSPC7110-100GEA
9720
CZK
APOSPC7110-100G
APOSPC7110-25G
Triethyloxonium tetrafluoroborate 95%
Triethyloxonium tetrafluoroborate
Powerful alkylating reagent for many reactions.
Vzorec:
C₆H₁₅BF₄O MW: 189,99 g/mol Bod tání: 84…89 °C Hustota: 1,328 g/cm³ (25 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00044423 CAS číslo: 368-39-8 EINECS: 206-705-1 UN: 2925 ADR: 4.1+8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...