Tisk…

3-Chloro-2-chloromethyl-5-(trifluoromethyl)pyridine 95%

Dodavatel: Apollo Scientific
Nebezpečí

PC7102-1G PC7102-5G
APOSPC7102-1GEA 7380 CZK
APOSPC7102-1G APOSPC7102-5G
3-Chloro-2-chloromethyl-5-(trifluoromethyl)pyridine 95%
3-Chloro-2-chloromethyl-5-(trifluoromethyl)pyridine
Vzorec: C₇H₄Cl₂F₃N
MW: 230,02 g/mol
Bod varu: 82…85 °C/15mmHg
Storage Temperature: Ambient
MDL Number: MFCD00173852
CAS číslo: 175277-52-8
UN: 3265
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR