Tisk…

Scandium(III) trifluoromethanesulphonate 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: Scandium(III) trifluoromethanesulphonate, Scandium triflate

PC7096-1G PC7096-5G PC7096-25G
APOSPC7096-1GEA 622 CZK
APOSPC7096-1G APOSPC7096-5G APOSPC7096-25G
Scandium(III) trifluoromethanesulphonate 97%
Scandium(III) trifluoromethanesulphonate
Vzorec: C₃F₉O₉S₃Sc
MW: 492,17 g/mol
Bod tání: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00192433
CAS číslo: 144026-79-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR