Tisk…

2-(2,2,2-Trifluoroethoxy)phenylboronic acid 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: (2-(2,2,2-Trifluoroethoxy)phenyl)boronic acid, 2-(2,2,2-Trifluoroethoxy)benzeneboronic acid

PC7093-1G PC7093-5G
APOSPC7093-1GEA 4860 CZK
APOSPC7093-1G APOSPC7093-5G
2-(2,2,2-Trifluoroethoxy)phenylboronic acid 98%
2-(2,2,2-Trifluoroethoxy)phenylboronic acid
Vzorec: C₈H₈BF₃O₃
MW: 219,95 g/mol
Bod tání: 125…128 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09027258
CAS číslo: 957060-90-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR