Tisk…

3-(2-Fluorophenylcarbamoyl)phenylboronic acid 98%

Dodavatel: Apollo Scientific
Varování

PC7091-1G PC7091-25G PC7091-5G
APOSPC7091-1GEA 834 CZK
APOSPC7091-1G APOSPC7091-25G APOSPC7091-5G
3-(2-Fluorophenylcarbamoyl)phenylboronic acid 98%
3-(2-Fluorophenylcarbamoyl)phenylboronic acid
Vzorec: C₁₃H₁₁BFNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD09027208
CAS číslo: 874288-35-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR