Tisk…

Potassium-2,4-bis(trifluoromethyl)phenyltrifluoroborate

Dodavatel: Apollo Scientific

PC7064-1G PC7064-5G
APOSPC7064-1GEA 4370 CZK
APOSPC7064-1G APOSPC7064-5G
Potassium-2,4-bis(trifluoromethyl)phenyltrifluoroborate
Potassium-2,4-bis(trifluoromethyl)phenyltrifluoroborate
Vzorec: C₈H₃BF₉K
Storage Temperature: Ambient
MDL Number: MFCD07368252
CAS číslo: 1150655-10-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR