Tisk…

5-Fluoro-3-pyridineboronic acid pinacol ester

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-Fluoropyridine-5-boronic acid pinacol ester, 3-Fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

PC7054-5G
APOSPC7054-5GEA 4400 CZK
APOSPC7054-5G
5-Fluoro-3-pyridineboronic acid pinacol ester
5-Fluoro-3-pyridineboronic acid pinacol ester
Vzorec: C₁₁H₁₅BFNO₂
MW: 223,05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07780754
CAS číslo: 719268-92-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR