2-Fluoro-5-methyl-3-pyridineboronic acid pinacol ester
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-Fluoro-5-methylpyridine-3-boronic acid pinacol ester, 2-Fluoro-5-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
PC7048-1G
PC7048-5G
APOSPC7048-1GEA
4670
CZK
APOSPC7048-1G
APOSPC7048-5G
2-Fluoro-5-methyl-3-pyridineboronic acid pinacol ester
2-Fluoro-5-methyl-3-pyridineboronic acid pinacol ester
Vzorec:
C₁₂H₁₇BFNO₂ MW: 237.0783 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08063092 CAS číslo: 1073371-96-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...