3-Fluoro-5-formylpyridine
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
5-Fluoronicotinaldehyde
PC7041-1G
PC7041-250MG
PC7041-5G
APOSPC7041-1GEA
3580
CZK
APOSPC7041-1G
APOSPC7041-250MG
APOSPC7041-5G
3-Fluoro-5-formylpyridine
3-Fluoro-5-formylpyridine
Vzorec:
C₆H₄FNO MW: 125,1 g/mol Bod varu: 71…76 °C/10mmHg Hustota: 1,27 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD07778429 CAS číslo: 39891-04-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...