Tisk…

2-Fluoro-5-formylpyridine 97%

Dodavatel: Apollo Scientific
Varování

PC7037-1G PC7037-5G PC7037-10G
APOSPC7037-1GEA 744 CZK
APOSPC7037-1G APOSPC7037-5G APOSPC7037-10G
2-Fluoro-5-formylpyridine 97%
2-Fluoro-5-formylpyridine
Vzorec: C₆H₄FNO
Storage Temperature: Ambient
MDL Number: MFCD08064049
CAS číslo: 677728-92-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR