Tisk…

2,5-Difluoro-4-methoxyphenylboronic acid 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: (2,5-Difluoro-4-methoxyphenyl)boronic acid

PC7026-1G PC7026-5G
APOSPC7026-1GEA 6850 CZK
APOSPC7026-1G APOSPC7026-5G
2,5-Difluoro-4-methoxyphenylboronic acid 98%
2,5-Difluoro-4-methoxyphenylboronic acid
Vzorec: C₇H₇BF₂O₃
MW: 187,94 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04038754
CAS číslo: 897958-93-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR