2,3-Difluoro-4-methoxyphenylboronic acid
Dodavatel: Apollo Scientific
|
Varování
|
PC7024-5G
PC7024-25G
APOSPC7024-5GEA
2030
CZK
APOSPC7024-5G
APOSPC7024-25G
2,3-Difluoro-4-methoxyphenylboronic acid
2,3-Difluoro-4-methoxyphenylboronic acid
Vzorec:
C₇H₇BF₂O₃ Storage Temperature: Refrigerator |
MDL Number:
MFCD07368234 CAS číslo: 170981-41-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...