Tisk…

4-Chloro-3-(trifluoromethyl)phenylboronic acid pinacol ester 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-(4-Chloro-3-(trifluoromethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

PC7018-5G
APOSPC7018-5GEA 1620 CZK
APOSPC7018-5G
4-Chloro-3-(trifluoromethyl)phenylboronic acid pinacol ester 97%
4-Chloro-3-(trifluoromethyl)phenylboronic acid pinacol ester
Vzorec: C₁₃H₁₅BClF₃O₂
MW: 306,516 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06657823
CAS číslo: 445303-09-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR