Tisk…

2-Chloro-6-(trifluoromethyl)phenylboronic acid

Dodavatel: Apollo Scientific
Varování

PC7017-1G PC7017-500MG
APOSPC7017-1GEA 8100 CZK
APOSPC7017-1G APOSPC7017-500MG
2-Chloro-6-(trifluoromethyl)phenylboronic acid
2-Chloro-6-(trifluoromethyl)phenylboronic acid
Vzorec: C₇H₅BClF₃O₂
MW: 224,3726 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09037968
CAS číslo: 851756-52-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR