Tisk…

5-Chloro-2-fluoro-4-methylphenylboronic acid

Dodavatel: Apollo Scientific

PC7015-1G PC7015-5G
APOSPC7015-1GEA 1130 CZK
APOSPC7015-1G APOSPC7015-5G
5-Chloro-2-fluoro-4-methylphenylboronic acid
5-Chloro-2-fluoro-4-methylphenylboronic acid
Vzorec: C₇H₇BClFO₂
Storage Temperature: Ambient
MDL Number: MFCD07368241
CAS číslo: 1072952-42-1

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR