Tisk…

5-Chloro-2,4-difluorophenylboronic acid pinacol ester

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-Chloro-2,4,-difluorophenylboronic acid pinacol ester, 2-(5-Chloro-2,4-difluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

PC7012-1G PC7012-5G
APOSPC7012-1GEA 2070 CZK
APOSPC7012-1G APOSPC7012-5G
5-Chloro-2,4-difluorophenylboronic acid pinacol ester
5-Chloro-2,4-difluorophenylboronic acid pinacol ester
Vzorec: C₁₂H₁₄BClF₂O₂
MW: 274,499 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08669564
CAS číslo: 1073354-65-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR