2-Bromo-5-fluoro-4-pyridineboronic acid 96%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(2-Bromo-5-fluoropyridin-4-yl)boronic acid, 2-Bromo-5-fluoropyridine-4-boronic acid, 2-Bromo-5-fluoro-4-pyridinylboronic acid
PC6997-1G
PC6997-5G
APOSPC6997-1GEA
6420
CZK
APOSPC6997-1G
APOSPC6997-5G
2-Bromo-5-fluoro-4-pyridineboronic acid 96%
2-Bromo-5-fluoro-4-pyridineboronic acid
Vzorec:
C₅H₄BBrFNO₂ MW: 219,8 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD04973353 CAS číslo: 1072951-43-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...