4-(3-Fluorophenylcarbamoyl)phenylboronic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-(3-Fluorophenylcarbamoyl)benzeneboronic acid, (4-((3-Fluorophenyl)carbamoyl)phenyl)boronic acid
PC6987-1G
PC6987-25G
PC6987-5G
APOSPC6987-1GEA
3740
CZK
APOSPC6987-1G
APOSPC6987-25G
APOSPC6987-5G
4-(3-Fluorophenylcarbamoyl)phenylboronic acid 98%
4-(3-Fluorophenylcarbamoyl)phenylboronic acid
Vzorec:
C₁₃H₁₁BFNO₃ MW: 259,04 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD09027204 CAS číslo: 874288-05-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...