3-(Trifluoromethoxy)toluene 98%
Dodavatel: Apollo Scientific
|
Varování
|
PC6982-25G
PC6982-5G
APOSPC6982-25GEA
4300
CZK
APOSPC6982-25G
APOSPC6982-5G
3-(Trifluoromethoxy)toluene 98%
3-(Trifluoromethoxy)toluene
Vzorec:
C₈H₇F₃O Storage Temperature: Ambient |
MDL Number:
MFCD07366526 CAS číslo: 705-44-2 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...