Tisk…

4-Fluoro-3-methoxyphenylacetic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-(4-fluoro-3-methoxyphenyl)acetic acid

PC6976-10G PC6976-1G PC6976-5G
APOSPC6976-10GEA 12870 CZK
APOSPC6976-10G APOSPC6976-1G APOSPC6976-5G
4-Fluoro-3-methoxyphenylacetic acid
4-Fluoro-3-methoxyphenylacetic acid
Vzorec: C₉H₉FO₃
MW: 184,166 g/mol
Bod tání: 103…107 °C
Storage Temperature: Ambient
MDL Number: MFCD08146650
CAS číslo: 946713-86-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR