Tetramethylammonium tetrafluoroborate 97%
Dodavatel: Apollo Scientific
PC6938-100G
PC6938-25G
PC6938-10G
PC6938-5G
APOSPC6938-100GEA
0
CZK
APOSPC6938-100G
APOSPC6938-25G
APOSPC6938-10G
APOSPC6938-5G
Tetramethylammonium tetrafluoroborate 97%
Tetramethylammonium tetrafluoroborate
Vzorec:
C₄H₁₂BF₄N Bod tání: 417…419 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011745 CAS číslo: 661-36-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...