3-Methyl-5-(trifluoromethoxy)benzonitrile 98%
Dodavatel: Apollo Scientific
PC6928-10G
PC6928-5G
PC6928-1G
APOSPC6928-10GEA
9990
CZK
APOSPC6928-10G
APOSPC6928-5G
APOSPC6928-1G
3-Methyl-5-(trifluoromethoxy)benzonitrile 98%
3-Methyl-5-(trifluoromethoxy)benzonitrile
Vzorec:
C₉H₆F₃NO MW: 201,15 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09025378 CAS číslo: 916420-59-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...