Tisk…

3-Chloro-5-(trifluoromethyl)benzonitrile

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 3-Chloro-5-cyanobenzotrifluoride

PC6915-1G PC6915-25G PC6915-5G
APOSPC6915-1GEA 622 CZK
APOSPC6915-1G APOSPC6915-25G APOSPC6915-5G
3-Chloro-5-(trifluoromethyl)benzonitrile
3-Chloro-5-(trifluoromethyl)benzonitrile
Vzorec: C₈H₃ClF₃N
Storage Temperature: Ambient
MDL Number: MFCD04115854
CAS číslo: 693245-52-2
UN: 3276
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR