Tisk…

Tetrafluorotetracyanoquinodimethane 97%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 2,3,5,6-Tetrafluoro-7,7,8,8-tetracyanoquinodimethane, TCNQF4, 2,2'-(Perfluorocyclohexa-2,5-diene-1,4-diylidene)dimalononitrile

PC6914-100MG PC6914-500MG
APOSPC6914-100MGEA 4170 CZK
APOSPC6914-100MG APOSPC6914-500MG
Tetrafluorotetracyanoquinodimethane 97%
Tetrafluorotetracyanoquinodimethane

Organic electronic material.

Vzorec: C₁₂F₄N₄
MW: 276,15 g/mol
Bod tání: 285…290 °C
Storage Temperature: Ambient
MDL Number: MFCD00042382
CAS číslo: 29261-33-4
UN: 3439
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR