Tetrafluoroterephthalonitrile 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
Perfluoroterephthalonitrile
PC6910-25G
PC6910-100G
PC6910-5G
APOSPC6910-25GEA
1580
CZK
APOSPC6910-25G
APOSPC6910-100G
APOSPC6910-5G
Tetrafluoroterephthalonitrile 98%
Tetrafluoroterephthalonitrile
Vzorec:
C₈F₄N₂ MW: 200,1 g/mol Bod tání: 197…199 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001776 CAS číslo: 1835-49-0 EINECS: 217-397-3 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...