Tetrafluoroterephthalic acid 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2,3,5,6-Tetrafluoroterephthalic acid
PC6900-1G
PC6900-5G
APOSPC6900-1GEA
622
CZK
APOSPC6900-1G
APOSPC6900-5G
Tetrafluoroterephthalic acid 97%
Tetrafluoroterephthalic acid
Vzorec:
C₈H₂F₄O₄ MW: 238,0900 g/mol Bod tání: 275…277 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00012301 CAS číslo: 652-36-8 EINECS: 211-489-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...