Tetrafluorosuccinyl chloride 95%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC6894-1G
PC6894-5G
APOSPC6894-1GEA
4730
CZK
APOSPC6894-1G
APOSPC6894-5G
Tetrafluorosuccinyl chloride 95%
Tetrafluorosuccinyl chloride
Vzorec:
C₄Cl₂F₄O₂ Bod varu: 85…87 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013653 CAS číslo: 356-15-0 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...