Tetrafluorosuccinic acid 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
2,2,3,3-Tetrafluorosuccinic acid, Tetrafluoro-1,4-butanedioic acid
PC6880-25G
PC6880-5G
APOSPC6880-25GEA
8670
CZK
APOSPC6880-25G
APOSPC6880-5G
Tetrafluorosuccinic acid 98%
Tetrafluorosuccinic acid
Vzorec:
C₄H₂F₄O₄ MW: 190,05 g/mol Bod varu: 150 °C/15mmHg Bod tání: 115…117 °C Hustota: 1,4 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00042430 CAS číslo: 377-38-8 EINECS: 206-820-7 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...