4-Bromo-7-fluoroindole
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-Bromo-7-fluoro-1H-indole
PC6872-1G
PC6872-250MG
APOSPC6872-1GEA
14920
CZK
APOSPC6872-1G
APOSPC6872-250MG
4-Bromo-7-fluoroindole
4-Bromo-7-fluoroindole
Vzorec:
C₈H₅BrFN MW: 214.04 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD03095318 CAS číslo: 883500-66-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...