2,2,3,3-Tetrafluoropropyl acrylate 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Acrylic Acid 2,2,3,3-Tetrafluoropropyl Ester
PC6868-25G
APOSPC6868-25GEA
5590
CZK
APOSPC6868-25G
2,2,3,3-Tetrafluoropropyl acrylate 98%
2,2,3,3-Tetrafluoropropyl acrylate
Vzorec:
C₆H₆F₄O₂ MW: 186,11 g/mol Bod varu: 132…134 °C Hustota: 1,348 g/cm³ (25 °C) Bod vzplanutí: 45 °C Storage Temperature: Refrigerator |
CAS číslo:
7383-71-3 UN: 1993 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...