3H-Tetrafluoropropionyl chloride 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC6867E-25G
PC6867E-5G
APOSPC6867E-25GEA
22580
CZK
APOSPC6867E-25G
APOSPC6867E-5G
3H-Tetrafluoropropionyl chloride 97%
3H-Tetrafluoropropionyl chloride
Vzorec:
C₃HClF₄O MW: 164,49 g/mol Bod varu: 38…40 °C Hustota: 1,521 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00153278 CAS číslo: 663-73-0 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...