Tisk…

Tetrafluorophthalic acid 98%

Dodavatel: Apollo Scientific
Varování

PC6840-100G PC6840-500G
APOSPC6840-100GEA 4170 CZK
APOSPC6840-100G APOSPC6840-500G
Tetrafluorophthalic acid 98%
Tetrafluorophthalic acid
Vzorec: C₈H₂F₄O₄
MW: 238,1 g/mol
Bod tání: 152…154 °C
Storage Temperature: Ambient
MDL Number: MFCD00002407
CAS číslo: 652-03-9
EINECS: 211-483-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR