Tisk…

α-Bromo-3-(difluoromethoxy)toluene 98%

Dodavatel: Apollo Scientific
Nebezpečí

PC6823-500MG PC6823-1G PC6823-5G
APOSPC6823-500MGEA 622 CZK
APOSPC6823-500MG APOSPC6823-1G APOSPC6823-5G
α-Bromo-3-(difluoromethoxy)toluene 98%
α-Bromo-3-(difluoromethoxy)toluene
Vzorec: C₈H₇BrF₂O
Bod varu: 76 °C/1mmHg
Storage Temperature: Ambient
MDL Number: MFCD00236232
CAS číslo: 72768-95-7
UN: 3265
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR