Tisk…

Tris(2,2,2-trifluoroethyl)borate 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: Boric Acid Tris(2,2,2-trifluoroethyl) Ester, Tris(2,2,2-trifluoroethyl) borate

PC6808-1G PC6808-25G PC6808-5G
APOSPC6808-1GEA 827 CZK
APOSPC6808-1G APOSPC6808-25G APOSPC6808-5G
Tris(2,2,2-trifluoroethyl)borate 97%
Tris(2,2,2-trifluoroethyl)borate
Vzorec: C₆H₆BF₉O₃
MW: 307.91 g/mol
Bod varu: 77 °C/200mmHg
Storage Temperature: Refrigerator
MDL Number: MFCD00093768
CAS číslo: 659-18-7
UN: 1993
ADR: 3 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR