N-Methyl-N-methoxydifluoroacetamide 95%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2,2-Difluoro-N-methoxy-N-methylacetamide
PC6769-1G
PC6769-5G
APOSPC6769-1GEA
827
CZK
APOSPC6769-1G
APOSPC6769-5G
N-Methyl-N-methoxydifluoroacetamide 95%
N-Methyl-N-methoxydifluoroacetamide
Vzorec:
C₄H₇F₂NO₂ MW: 139,1 g/mol Bod varu: 40…41 °C/0,7mmHg Storage Temperature: Ambient |
MDL Number:
MFCD06248152 CAS číslo: 142492-01-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...