Tisk…

1,2-Dibromopentafluoropropyl-2,2,3,3-tetrafluoropropyl ether 97%

Dodavatel: Apollo Scientific

PC6752-25G PC6752-5G
APOSPC6752-25GEA 12570 CZK
APOSPC6752-25G APOSPC6752-5G
1,2-Dibromopentafluoropropyl-2,2,3,3-tetrafluoropropyl ether 97%
1,2-Dibromopentafluoropropyl-2,2,3,3-tetrafluoropropyl ether
Vzorec: C₆H₃Br₂F₉O
Storage Temperature: Ambient
MDL Number: MFCD03094408
CAS číslo: 396716-51-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR