4-Trifluoromethoxythioanisole 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Methyl(4-(trifluoromethoxy)phenyl)sulphane, 4-(Trifluoromethoxy)phenyl methyl sulphphide, 4-(Trifluoromethoxy)phenyl methyl sulphide
PC6743-5G
PC6743-25G
APOSPC6743-5GEA
3770
CZK
APOSPC6743-5G
APOSPC6743-25G
4-Trifluoromethoxythioanisole 98%
4-Trifluoromethoxythioanisole
Vzorec:
C₈H₇F₃OS MW: 208,20 g/mol Bod varu: 112…114 °C/18mmHg Storage Temperature: Ambient |
MDL Number:
MFCD01320776 CAS číslo: 2546-45-4 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...