2,3,4,5-Tetrafluorobenzoyl chloride 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
2,3,4,5-tetrafluorobenzoylchloride
PC6742-100G
PC6742-250G
PC6742-25G
PC6742-500G
APOSPC6742-100GEA
2650
CZK
APOSPC6742-100G
APOSPC6742-250G
APOSPC6742-25G
APOSPC6742-500G
2,3,4,5-Tetrafluorobenzoyl chloride 98%
2,3,4,5-Tetrafluorobenzoyl chloride
Vzorec:
C₇HClF₄O MW: 212,53 g/mol Hustota: 1,58 g/cm³ (25 °C) Bod vzplanutí: 90 °C Storage Temperature: Ambient |
MDL Number:
MFCD00075164 CAS číslo: 94695-48-4 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...