Tetrafluoro-p-benzoquinone 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Tetrafluoroquinone, 2,3,5,6-Tetrafluoro-1,4-benzoquinone, Tetrafluoro-1,4-benzoquinone, p-Fluoranil, 4-Fluoranil
PC6740-1G
APOSPC6740-1GEA
1040
CZK
APOSPC6740-1G
Tetrafluoro-p-benzoquinone 97%
Tetrafluoro-p-benzoquinone
High potential quinone; undergoes Diels-Alder cyclo-additions; complexes with a wide range of electron donors
Vzorec:
C₆F₄O₂ MW: 180,06 g/mol Bod tání: 183…186 °C (subl.) Storage Temperature: Ambient |
MDL Number:
MFCD00001592 CAS číslo: 527-21-9 EINECS: 208-411-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...