3-(4-Fluorophenyl)-1H-pyrazole-4-carbaldehyde 97%
Dodavatel: Apollo Scientific
Synonyms:
3-(4-Fluorophenyl)pyrazole-4-carboxaldehyde
PC6717-25G
PC6717-5G
APOSPC6717-25GEA
5790
CZK
APOSPC6717-25G
APOSPC6717-5G
3-(4-Fluorophenyl)-1H-pyrazole-4-carbaldehyde 97%
3-(4-Fluorophenyl)-1H-pyrazole-4-carbaldehyde
Vzorec:
C₁₀H₇FN₂O MW: 190,18 g/mol Bod tání: 166…167 °C Storage Temperature: Ambient |
MDL Number:
MFCD01922131 CAS číslo: 306936-57-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...