Tisk…

2,4-Dibromo-6-fluorophenol 97%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 1,3-Dibromo-5-fluoro-4-hydroxybenzene

PC6713-100G PC6713-25G PC6713-5G
APOSPC6713-100GEA 5390 CZK
APOSPC6713-100G APOSPC6713-25G APOSPC6713-5G
2,4-Dibromo-6-fluorophenol 97%
2,4-Dibromo-6-fluorophenol
Vzorec: C₆H₃Br₂FO
MW: 269,9 g/mol
Bod tání: 35…37 °C
Storage Temperature: Ambient
MDL Number: MFCD00042233
CAS číslo: 576-86-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR