Tisk…

1H,1H-Undecafluorohexylamine 97%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 1H,1H-Perfluorohexylamine

PC6703-1G PC6703-5G
APOSPC6703-1GEA 10580 CZK
APOSPC6703-1G APOSPC6703-5G
1H,1H-Undecafluorohexylamine 97%
1H,1H-Undecafluorohexylamine
Vzorec: C₆H₄F₁₁N
MW: 299,09 g/mol
Bod varu: 107 °C/740mmHg
Storage Temperature: Ambient
MDL Number: MFCD09025929
CAS číslo: 355-34-0
UN: 2735
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR