Tisk…

2-Fluoro-m-cresol

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Fluoro-3-methylphenol

PC6668-5G PC6668-1G PC6668-25G
APOSPC6668-5GEA 910 CZK
APOSPC6668-5G APOSPC6668-1G APOSPC6668-25G
2-Fluoro-m-cresol
2-Fluoro-m-cresol
Vzorec: C₇H₇FO
MW: 126,1300 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09835186
CAS číslo: 77772-72-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR