Tetraethylammonium fluoride hydrate 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Tetrylammonium fluoride hydrate, Tetraethylammonium fluoride xhydrate
PC6665-100G
PC6665-25G
PC6665-500G
APOSPC6665-100GEA
1910
CZK
APOSPC6665-100G
APOSPC6665-25G
APOSPC6665-500G
Tetraethylammonium fluoride hydrate 98%
Tetraethylammonium fluoride hydrate
Vzorec:
C₈H₂₀FN·xH₂O MW: 167.27 Bod tání: 120…121 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149990 CAS číslo: 98330-04-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...