Perfluoroazelaic acid, Technical Grade
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
Perfluorononane-1,9-dioic acid
PC6656-5G
PC6656-1G
APOSPC6656-5GEA
2900
CZK
APOSPC6656-5G
APOSPC6656-1G
Perfluoroazelaic acid, Technical Grade
Perfluoroazelaic acid
Vzorec:
HO₂C(CF₂)₇CO₂H Bod varu: 215 °C/60mmHg Storage Temperature: Ambient |
MDL Number:
MFCD00153274 CAS číslo: 23453-64-7 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...